Learn More
TRIM58 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Product Code. 18405755 Shop All Bio Techne Products
Product Code. 18405755 Supplier Novus Biologicals Supplier No. NBP281870MFV450
Rabbit Polyclonal Antibody
TRIM58 Polyclonal antibody specifically detects TRIM58 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA