missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM58 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81870
This item is not returnable.
View return policy
Description
TRIM58 Polyclonal specifically detects TRIM58 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| TRIM58 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| TRIM58 | |
| TRIM58 antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TRIM58. Amino Acid Squence: DTLPRKGETTPSPENGV | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 ug/mL, ELISA | |
| BIA2, DKFZp434C091, Protein BIA2, tripartite motif containing 58, tripartite motif-containing 58, tripartite motif-containing protein 58 | |
| Rabbit | |
| 56 kDa | |
| 0.1 mg | |
| Zinc Finger | |
| 25893 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction