missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM56 Antibody (OTI5A1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74613JF646
This item is not returnable.
View return policy
Description
TRIM56 Monoclonal specifically detects TRIM56 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TRIM56 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TRIM56 | |
| Recombinant protein fragment corresponding to amino acids 311-554 of human TRIM56 (NP_112223) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI5A1 | |
| Western Blot, Immunohistochemistry | |
| DKFZp667O116, E3 ubiquitin-protein ligase TRIM56, EC 6.3.2.-, RING finger protein 109, RNF109FLJ35608, tripartite motif containing 56, tripartite motif-containing 56, Tripartite motif-containing protein 56 | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 81844.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction