missing translation for 'onlineSavingsMsg'
Learn More

TRIM5 gamma Antibody, DyLight 405, Novus Biologicals™

Product Code. 18432586 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432586 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432586 Supplier Novus Biologicals Supplier No. NBP176604V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIM5 gamma Polyclonal antibody specifically detects TRIM5 gamma in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM5 gamma
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of human TRIM5 gamma. The immunogen is located within the last 50 amino acids of TRIM5 gamma. Amino Acid Squence: FKIYDSPSKTHITY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Lipid and Metabolism, Virology Bacteria and Parasites, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 85363
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.