missing translation for 'onlineSavingsMsg'
Learn More

TRIM5 delta Polyclonal Antibody
GREENER_CHOICE

Product Code. 12848262
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848262 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848262 Supplier Invitrogen Supplier No. PA520048

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TRIM5 delta Polyclonal Antibody

TRIM5 delta Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM5 delta
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TRIM5
Gene Accession No. Q9C035
Gene Alias EG667823; Gm8833; RING finger protein 88; RING-type E3 ubiquitin transferase TRIM5; RNF88; TRIM5; TRIM5alpha; tripartite motif containing 5; tripartite motif containing 5 transcript variant iota; tripartite motif containing 5 transcript variant kappa; tripartite motif protein TRIM5; tripartite motif-containing 5; tripartite motif-containing protein 5
Gene Symbols TRIM5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 13 amino acids near the carboxy terminus of human TRIM5 delta.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 667823, 85363
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.