missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM5 alpha Polyclonal Antibody

Product Code. 12868262
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12868262 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12868262 Supplier Invitrogen™ Supplier No. PA520050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~55kDa. A suggested positive control is human stomach tissue lysate. PA5-20050 can be used with blocking peptide PEP-0169.

The protein encoded by this gene is a member of the tripartite motif family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein forms homo-oligomers via the coiled-coil region and localizes to cytoplasmic bodies. It appears to function as a E3 ubiquitin-ligase and ubiquitinates itself to regulate its subcellular localization. It may play a role in retroviral restriction. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM5 alpha
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Trim5
Gene Accession No. Q0PF16, Q9C035
Gene Alias EG667823; Gm8833; HGNC:16276; OTTHUMP00000069812; RING finger protein 88; RING-type E3 ubiquitin transferase TRIM5; RNF88; TRIM5; Trim5 alpha; TRIM5 alpha; predicted E3 ubiquiting ligase; TRIM5 gamma; predicted E3 ubiquiting ligase; trim5a; TRIM5alpha; tripartite motif containing 5; tripartite motif containing 5 transcript variant iota; tripartite motif containing 5 transcript variant kappa; tripartite motif protein TRIM5; tripartite motif-containing 5; tripartite motif-containing 5 alpha isoform; tripartite motif-containing 5 gamma isoform; tripartite motif-containing 5/cyclophilin A fusion protein; tripartite motif-containing protein 5; tripartite motif-containing protein 5 isoform alpha
Gene Symbols Trim5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids near the C-terminus of rhesus monkey TRIM5 alpha.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 574288, 85363
Target Species Human, Rhesus Macaque
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.