Learn More
Invitrogen™ TRIM5 alpha Monoclonal Antibody (K94032_3D8), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | TRIM5 alpha |
|---|---|
| Clone | K94032_3D8 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30196847
|
Invitrogen™
MA556083 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30206168
|
Invitrogen™
MA555703 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: GHSFCQACLT ANHKKSMLDK GESSCPVCRI SYQPENIRPN RHVANIVEKL REVKLSPEGQ KVDHCARHGE KLLLFCQEDG KVICWLCERS QEHRGHHTFL TEEVAREYQV KLQAALEMLR QKQQEAEELE ADIREEKASW KTQIQYDKTN VLADFEQLRD ILDWEESNEL QNLEKEEEDI LKSLTNSETE MVQQTQSLRE LISDLEHRLQ GSVMELLQGV DGVIKRTENV TLKKPETFPK
TRIM5 alpha Monoclonal antibody specifically detects TRIM5 alpha in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| TRIM5 alpha | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| EG667823; Gm8833; HGNC:16276; OTTHUMP00000069812; RING finger protein 88; RING-type E3 ubiquitin transferase TRIM5; RNF88; TRIM5; Trim5 alpha; TRIM5 alpha; predicted E3 ubiquiting ligase; TRIM5 gamma; predicted E3 ubiquiting ligase; trim5a; TRIM5alpha; tripartite motif containing 5; tripartite motif containing 5 transcript variant iota; tripartite motif containing 5 transcript variant kappa; tripartite motif protein TRIM5; tripartite motif-containing 5; tripartite motif-containing 5 alpha isoform; tripartite motif-containing 5 gamma isoform; tripartite motif-containing 5/cyclophilin A fusion protein; tripartite motif-containing protein 5; tripartite motif-containing protein 5 isoform alpha | |
| Recombinant human TRIM5 (immunogen range is 31-270). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Trim5 |
| K94032_3D8 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9C035 | |
| 85363 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.