missing translation for 'onlineSavingsMsg'
Learn More

TRIM5 alpha Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18404678 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404678 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404678 Supplier Novus Biologicals Supplier No. NBP176600JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIM5 alpha Polyclonal antibody specifically detects TRIM5 alpha in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM5 alpha
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to amino acids near the C-terminus of rhesus monkey TRIM5 alpha. The immunogen is located within the last 50 amino acids of TRIM5 alpha. Amino Acid Squence: SFSKPVFPYLNPRK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Lipid and Metabolism, Virology Bacteria and Parasites, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 85363
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.