missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM5 alpha Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-76600-0.025mg
This item is not returnable.
View return policy
Description
TRIM5 alpha Polyclonal specifically detects TRIM5 alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| TRIM5 alpha | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5 | |
| Rabbit | |
| 55 kDa | |
| 0.025 mg | |
| Immunology, Lipid and Metabolism, Virology Bacteria and Parasites, Zinc Finger | |
| 85363 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| NP_149023 | |
| TRIM5 | |
| Antibody was raised against a synthetic peptide corresponding to amino acids near the C-terminus of rhesus monkey TRIM5 alpha. The immunogen is located within the last 50 amino acids of TRIM5 alpha. Amino Acid Squence: SFSKPVFPYLNPRK | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Anti-TRIM5 alpha is human specific but should presumably also react with rhesus protein. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction