missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM47 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550892
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total TRIM47 protein.
E3 ubiquitin-protein ligase that mediates the ubiquitination and proteasomal degradation of CYLD. [UniProt].
Specifications
| TRIM47 | |
| Polyclonal | |
| Unconjugated | |
| TRIM47 | |
| 2210023F24Rik; E3 ubiquitin-protein ligase TRIM47; Gene overexpressed in astrocytoma protein; GOA; RING finger protein 100; RNF100; Trim47; tripartite motif containing 47; tripartite motif protein 47; tripartite motif-containing 47; tripartite motif-containing protein 47 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 217333, 91107 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.9 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8C0E3, Q96LD4 | |
| TRIM47 | |
| Synthetic peptide corresponding to a region derived from internal residues of human tripartite motif containing 47. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction