missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM44 Antibody (OTI1E10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74611CL1
This item is not returnable.
View return policy
Description
TRIM44 Monoclonal antibody specifically detects TRIM44 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| TRIM44 | |
| Monoclonal | |
| CoraFluor 1 | |
| DIPBMGC3490, HSA249128, MC7, Protein DIPB, tripartite motif containing 44, tripartite motif-containing 44, tripartite motif-containing protein 44 | |
| Full length human recombinant protein of human TRIM44(NP_060053) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI1E10 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 54765 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction