missing translation for 'onlineSavingsMsg'
Learn More

TRIM41 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18341963 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341963 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341963 Supplier Novus Biologicals Supplier No. NBP305293AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIM41 Polyclonal antibody specifically detects TRIM41 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM41
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias tripartite motif containing 41
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of the human TRIM41 protein (between amino acids 400-470) [UniProt Q8WV44].
Purification Method Immunogen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 90933
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.