missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110979
This item is not returnable.
View return policy
Description
Immunogen sequence: VAVGVCREDV MGITDRSKMS PDVGIWAIYW SAAGYWPLIG FPGTPTQQEP ALHRVGVYLD RGTGNVSFYS AVDGVHLHTF SCSSVSRLRP FFWLSPLASL VIPPV Highest antigen sequence indentity to the following orthologs: Rat - 38%, Mouse - 38%.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternative splicing of this gene generates two transcript variants.
Specifications
| TRIM4 | |
| Polyclonal | |
| Unconjugated | |
| TRIM4 | |
| E3 ubiquitin-protein ligase TRIM4; RING finger protein 87; RING-type E3 ubiquitin transferase TRIM4; RNF87; TRIM4; tripartite motif containing 4; tripartite motif protein 4; tripartite motif-containing 4; Tripartite motif-containing protein 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 89122 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C037 | |
| TRIM4 | |
| Recombinant Protein corresonding to Human TRIM4. Recombinant protein control fragment (Product #RP-92755). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction