missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80824
This item is not returnable.
View return policy
Description
TRIM4 Polyclonal specifically detects TRIM4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TRIM4 | |
| Polyclonal | |
| Western Blot 1:100-1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Q9C037 | |
| TRIM4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KSQRNLVAKMKKVMHLQDVEVKNATQWKDKIKSQRMRISTEFSKLHNFLVEEEDLFLQRLNKEEEETKKKLNENTLKLNQTIASLKKLILEVGEKSQAPTLELLQNPK | |
| 0.1 mL | |
| Zinc Finger | |
| 89122 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RING finger protein 87, RNF87tripartite motif protein 4, tripartite motif containing 4, tripartite motif-containing 4, tripartite motif-containing protein 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction