missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM38 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540973
This item is not returnable.
View return policy
Description
Peptide sequence: SPAVCAAVTA LQNYLTEALK AMDKMYLSNN PNSHTDNNAK SSDKEEKHRK Sequence homology: Guinea Pig: 90%; Human: 100%.
This gene encodes a member of the tripartite motif (TRIM) family. The encoded protein contains a RING-type zinc finger, B box-type zinc finger and SPRY domain. The function of this protein has not been identified. A pseudogene of this gene is located on the long arm of chromosome 4.
Specifications
| TRIM38 | |
| Polyclonal | |
| Unconjugated | |
| TRIM38 | |
| E3 ubiquitin-protein ligase TRIM38; Gm23; hypothetical protein LOC531022; MGC8946; RING finger protein 15; RING-type E3 ubiquitin transferase TRIM38; RNF15; Ro/SSA ribonucleoprotein homolog; RORET; TRIM38; tripartite motif containing 38; tripartite motif-containing 38; tripartite motif-containing protein 38; tripartite motif-containing protein 38; E3 ubiquitin-protein ligase TRIM38; tripartite motif-containing protein 38; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM38; zinc finger protein RoRet | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10475 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00635 | |
| TRIM38 | |
| Synthetic peptide directed towards the middle region of human TRIM38. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction