missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM38 Antibody (OTI113D3), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74610IR
This item is not returnable.
View return policy
Description
TRIM38 Monoclonal specifically detects TRIM38 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TRIM38 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TRIM38 | |
| Recombinant protein fragment corresponding to amino acids 1-265 of human TRIM38 (NP_006346) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI113D3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| MGC8946, RING finger protein 15RORETRo/SSA ribonucleoprotein homolog, RNF15, tripartite motif containing 38, tripartite motif-containing 38, tripartite motif-containing protein 38, Zinc finger protein RoRet, zinc-finger protein RoRet | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction, Zinc Finger | |
| 10475.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction