missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM38 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93955-0.02ml
This item is not returnable.
View return policy
Description
TRIM38 Polyclonal antibody specifically detects TRIM38 in Human samples. It is validated for Western Blot
Specifications
| TRIM38 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| MGC8946, RING finger protein 15RORETRo/SSA ribonucleoprotein homolog, RNF15, tripartite motif containing 38, tripartite motif-containing 38, tripartite motif-containing protein 38, Zinc finger protein RoRet, zinc-finger protein RoRet | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 266-465 of human TRIM38 (NP_006346.1). VSLELHTMCNVSKLYFDVKKMLRSHQVSVTLDPDTAHHELILSEDRRQVTRGYTQENQDTSSRRFTAFPCVLGCEGFTSGRRYFEVDVGEGTGWDLGVCMENVQRGTGMKQEPQSGFWTLRLCKKKGYVALTSPPTSLHLHEQPLLVGIFLDYEAGVVSFYNGNTGCHIFTFPKASFSDTLRPYFQVYQYSPLFLPPPGD | |
| 0.02 mL | |
| Signal Transduction, Zinc Finger | |
| 10475 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction