missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM37 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA582779

Product Code. 16389734

  • £443.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IHLLDLKDRS SIENLWGLQP RPPASLLQPT ASYSRKDKDQ RKQQAMWRVP SDLKMLKRLK TQMAEVRCMK TDVKNTLSEI KSSSAASGDM QTSLFSADQA ALAACGTENS GRLQDLGMEL LAKSS Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.

Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRIM37
Polyclonal
Unconjugated
TRIM37
1110032A10Rik; 2810004E07Rik; AI848587; AU043018; E3 ubiquitin-protein ligase TRIM37; Kiaa0898; mKIAA0898; MUL; mulibrey nanism protein; POB1; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; TEF3; TRAF encompassing factor 3; Trim37; tripartite motif containing 37; tripartite motif protein 37; tripartite motif-containing 37; tripartite motif-containing protein 37
Rabbit
Antigen affinity chromatography
RUO
4591
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.20 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O94972
TRIM37
Recombinant protein corresponding to Human TRIM37. Recombinant protein control fragment (Product #RP-93190).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.