missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM37 Monoclonal Antibody (2D11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA519380
This item is not returnable.
View return policy
Description
Peptide Sequence: GHLEGLQMTD LENNSETGEL QPVLPEGASA APEEGMSSDS DIECDTENEE QEEHTSVGGF HDSFMVMTQP PDEDTHSSFP DGEQIGPEDL SFNTDENSGR.
Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
Specifications
| TRIM37 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O94972 | |
| TRIM37 | |
| TRIM37 (NP_056109, 865 a.a. ~ 964 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2D11 | |
| Unconjugated | |
| TRIM37 | |
| 1110032A10Rik; 2810004E07Rik; AI848587; AU043018; E3 ubiquitin-protein ligase TRIM37; Kiaa0898; mKIAA0898; MUL; mulibrey nanism protein; POB1; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; TEF3; TRAF encompassing factor 3; Trim37; tripartite motif containing 37; tripartite motif protein 37; tripartite motif-containing 37; tripartite motif-containing protein 37 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4591 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction