missing translation for 'onlineSavingsMsg'
Learn More

TRIM32 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15574862
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15574862 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574862 Supplier Invitrogen Supplier No. PA541535

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TRIM32 Polyclonal Antibody

TRIM32 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM32
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Trim32
Gene Accession No. Q13049
Gene Alias 1810045E12Rik; 3f3; 72 kDa Tat-interacting protein; BBS11; bM468K2.2 (tripartite motif protein 32); E3 ubiquitin-protein ligase TRIM32; HT2A; LGMD2H; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; TATIP; Trim32; tripartite motif containing 32; tripartite motif protein 32; tripartite motif-containing 32; tripartite motif-containing protein 32; Zfp117; zinc finger protein 117; zinc finger protein HT2A; zinc-finger protein HT2A
Gene Symbols Trim32
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TRIM32 (aa 559-608).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22954
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.