missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM32 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149214
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.
Specifications
| TRIM32 | |
| Polyclonal | |
| Unconjugated | |
| Trim32 | |
| 1810045E12Rik; 3f3; 72 kDa Tat-interacting protein; BBS11; bM468K2.2 (tripartite motif protein 32); E3 ubiquitin-protein ligase TRIM32; HT2A; LGMD2H; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; TATIP; Trim32; tripartite motif containing 32; tripartite motif protein 32; tripartite motif-containing 32; tripartite motif-containing protein 32; Zfp117; zinc finger protein 117; zinc finger protein HT2A; zinc-finger protein HT2A | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 22954, 69807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q13049, Q8CH72 | |
| Trim32 | |
| E. coli-derived recombinant human TRIM32, Arg105-Lys204. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction