missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM3/BERP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | TRIM3/BERP |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654086
|
Novus Biologicals
NBP2-48791-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643116
|
Novus Biologicals
NBP2-48791 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIM3/BERP Polyclonal antibody specifically detects TRIM3/BERP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TRIM3/BERP | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BERPRING finger protein 97, Brain-expressed RING finger protein, HAC1, RING finger protein 22, RNF22brain expressed ring finger, RNF97FLJ16135, tripartite motif containing 3, tripartite motif protein TRIM3, tripartite motif-containing 3, tripartite motif-containing protein 3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TQRQGQEHIGSSCSFAEQALRLGSAPEVLLVRKHMRERLAALAAQAFPERPHENAQLELVLEVDGLRRSVLNLGALLTTSATAHETVAT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 10612 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title