missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM28 Polyclonal Antibody

TRIM28 Polyclonal Antibody
Brand: Invitrogen PA534485
This item is not returnable.
View return policy
Description
TRIM28 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| TRIM28 | |
| Polyclonal | |
| Unconjugated | |
| TRIM28 | |
| AA408787; E3 SUMO-protein ligase TRIM28; KAP1; KAP-1; KRAB [Kruppel-associated box domain]-associated protein 1; KRAB-A-interacting protein; KRAB-associated protein 1; KRAB-interacting p; KRAB-interacting protein 1; Krip1; KRIP-1; MommeD9; Nuclear corepressor KAP-1; PPP1R157; protein phosphatase 1, regulatory subunit 157; RING finger protein 96; RING-type E3 ubiquitin transferase TIF1-beta; RNF96; TF1B; tif1 beta; Tif1b; Tif1beta; TIF1-beta; Transcription intermediary factor 1-beta; transcriptional intermediary factor 1, beta; transcriptional intermediary factor 1-beta; Trim28; tripartite motif; tripartite motif containing 28; tripartite motif protein 28; tripartite motif-containing 28; tripartite motif-containing protein 28 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10155, 116698, 21849 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O08629, Q13263, Q62318 | |
| TRIM28 | |
| A 14 amino acid peptide near the carboxy terminus of human TRIM28. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction