missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM28, Mouse anti-Human, Clone: 4E6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010155-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: IVDPVEPHGEMKFQWDLNAWTKSAEAFGKIVAERPGTNSTGPAPMAPPRAPGPLSKQGSGSSQPMEVQEGYGFGSGDDPYSSAEPHVSGVKRSRSGEGEVSGLMRKVPRVSLERLDLDLTADSQPPVFKVFPGSTTEDYNLIVIERSpecifications
| TRIM28 | |
| Monoclonal | |
| Unconjugated | |
| tripartite motif-containing 28 | |
| FLJ29029/KAP1/RNF96/TF1B/TIF1B | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown, Western Blot | |
| 4E6 | |
| In 1x PBS, pH 7.4 | |
| BC004978 | |
| TRIM28 | |
| TRIM28 (AAH04978, 379 a.a. ∼ 524 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10155 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction