missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM27 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567533
Additional Details : Weight : 0.01000kg
Description
This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the nuclear matrix. It interacts with the enhancer of polycomb protein and represses gene transcription. It is also thought to be involved in the differentiation of male germ cells. Fusion of the N-terminus of this protein with the truncated C-terminus of the RET gene product has been shown to result in production of the ret transforming protein.Specifications
TRIM27 | |
Polyclonal | |
Unconjugated | |
TRIM27 | |
AW538890; Gm19403; ret finger protein; ret finger protein 1; RFP; RFP transforming protein; Rfp1; RING finger protein 76; RING-type E3 ubiquitin transferase TRIM27; RNF76; Trim27; tripartite motif containing 27; tripartite motif protein 27; tripartite motif protein TRIM27; tripartite motif-containing 27; tripartite motif-containing protein 27; Zinc finger protein RFP | |
Rabbit | |
affinity chromatography | |
RUO | |
19720, 5987 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P14373, Q62158 | |
TRIM27 | |
Fusion protein of human TRIM27 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |