missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527603
This item is not returnable.
View return policy
Description
Recommended positive controls: K562, THP-1, HL-60, U87-MG, SK-N-SH, IMR32, rat testis. Predicted reactivity: Mouse (97%), Rat (96%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the nuclear matrix. It interacts with the enhancer of polycomb protein and represses gene transcription. It is also thought to be involved in the differentiation of male germ cells. Fusion of the N-terminus of this protein with the truncated C-terminus of the RET gene product has been shown to result in production of the ret transforming protein.
Specifications
| TRIM27 | |
| Polyclonal | |
| Unconjugated | |
| TRIM27 | |
| AW538890; Gm19403; ret finger protein; ret finger protein 1; RFP; RFP transforming protein; Rfp1; RING finger protein 76; RING-type E3 ubiquitin transferase TRIM27; RNF76; Trim27; tripartite motif containing 27; tripartite motif protein 27; tripartite motif protein TRIM27; tripartite motif-containing 27; tripartite motif-containing protein 27; Zinc finger protein RFP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19720, 291171, 5987 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.66 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P14373, Q62158 | |
| TRIM27 | |
| Recombinant protein encompassing a sequence within the center region of human TRIM27. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction