missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35634-100ul
This item is not returnable.
View return policy
Description
TRIM27 Polyclonal antibody specifically detects TRIM27 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| TRIM27 | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Ret finger proteinRING finger protein 76, RFPtripartite motif protein TRIM27, RNF76RFP transforming protein, tripartite motif containing 27, tripartite motif-containing 27, Tripartite motif-containing protein 27, zinc finger protein RFP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human TRIM27 (NP_006501.1).,, Sequence:, MASGSVAECLQQETTCPVCLQYFAEPMMLDCGHNICCACLARCWGTAETNVSCPQCRETFPQRHMRPNRHLANVTQLVKQLRTERPSGPGGEMGVCEKHREPLKLYCEEDQMPICVVCDRSREHRGHSVLPLEEAVEGFKEQIQNQLDHLKRVKDLKKRRRAQGEQARAELLSLTQMEREKIVWEFEQLYHSLKEHEYRL | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5987 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction