missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM26 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27013-1-AP-20UL
This item is not returnable.
View return policy
Description
TRIM26 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The gene localizes to the major histocompatibility complex (MHC) class I region on chromosome 6.Specifications
| TRIM26 | |
| Polyclonal | |
| Unconjugated | |
| TRIM26 | |
| Acid finger protein, AFP, RING finger protein 95, RNF95, TRIM26, tripartite motif containing 26, Zinc finger protein 173, ZNF173 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22670, 309586, 7726 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62603, Q12899, Q99PN3 | |
| TRIM26 | |
| TRIM26 Fusion Protein Ag25546 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction