missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM24 Recombinant Rabbit Monoclonal Antibody (K01_2P84)
Description
TRIM24 Recombinant Monoclonal Antibody for Western Blot
The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
Specifications
| Antigen | TRIM24 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2P84 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | TRIM24 |
| Gene Accession No. | O15164 |
| Gene Alias | A130082H20Rik; Ab2-427; AI447469; D430004I05Rik; E3 ubiquitin-protein ligase TRIM24; hTIF1; PTC6; RING finger protein 82; RING-type E3 ubiquitin transferase TIF1-alpha; RNF82; TF1A; Tif1; Tif1a; TIF1alpha; TIF1-alpha; Transcription intermediary factor 1-alpha; transcriptional intermediary factor 1; transcriptional intermediary factor 1, alpha; Trim24; tripartite motif containing 24; tripartite motif-containing 24; tripartite motif-containing protein 24 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?