missing translation for 'onlineSavingsMsg'
Learn More

TRIM24 Mouse anti-Human, Mouse, Clone: 1D6D9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66324-1-IG-20UL

 View more versions of this product

Product Code. 16858095

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc binding domains (a RING, a B box type 1 and a B box type 2) and a coiled coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications

Specifications

TRIM24
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15164, Q64127
TRIM24
TRIM24 Fusion Protein Ag5417
20 μL
Primary
Mouse, Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1D6D9
Unconjugated
TRIM24
hTIF1, PTC6, RING finger protein 82, RNF82, TF1A, TIF1, TIF1 alpha, TIF1A, TIF1ALPHA, TRIM24, tripartite motif containing 24
Mouse
Protein G
RUO
21848, 8805
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.