missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM23 Recombinant Rabbit Monoclonal Antibody (K01_2J56)

Product Code. 30828796
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828796 50 μL
30827908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828796 Supplier Invitrogen™ Supplier No. MA565352

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TRIM23 Recombinant Monoclonal Antibody for Western Blot

Acts as an E3 ubiquitin-protein ligase. In the presence of the human cytomegalovirus (HCMV) protein UL144, participates in 'Lys-63'-linked auto-ubiquitination of TRAF6 resulting in the virally controlled activation of NF-kappa-B at early time of infection. The C-terminus can act as an allosteric activator of the cholera toxin catalytic subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM23
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2J56
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene TRIM23
Gene Accession No. P36406, P36407, Q8BGX0
Gene Alias 6330516O20Rik; ADP-ribosylation factor domain protein 1; ADP-ribosylation factor domain protein 1 64kD; ADP-ribosylation factor domain protein 1, 64kD; ADP-ribosylation factor domain protein 1, 64kDa; ADP-ribosylation factor domain-containing protein 1; AI450195; ARD1; Ard-1; ARF domain protein 1; ARFD1; E3 ubiquitin-protein ligase TRIM23; GTP-binding protein ARD-1; nucleotide binding protein; RING finger protein 46; RING-type E3 ubiquitin transferase TRIM23; RNF46; TRIM23; tripartite motif containing 23; tripartite motif protein 23; tripartite motif protein TRIM23; tripartite motif-containing 23; tripartite motif-containing protein 23
Gene Symbols TRIM23
Host Species Rabbit
Immunogen A synthetic peptide of human TRIM23.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 373, 81002, 81003
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.