Learn More
Invitrogen™ TRIM23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597010
Description
Immunogen sequence: MATLVVNKLG AGVDSGRQGS RGTAVVKVLE CGVCEDVFSL QGDKVPRLLL CGHTVCHDCL TRLPLHGRAI RCPFDRQVTD LGDSGVWGLK KNFALLELLE RLQNGPIGQY GAAEESIGIS GESIIRCDED EAHLASVYCT VCATHLCSEC SQVTHSTKTL AKHRRVPLAD KPHEKTMCSQ HQVHAIEFVC LEEGCQTSPL MCCVCKEYGK HQGHKHSVLE PEANQIRASI LDMAHCIRTF TEEISDYSRK LVGIVQHIEG GEQIVEDGIG MAHTEHVPGT; Positive Samples: PC-12, Mouse spleen, Mouse brain; Cellular Location: Endomembrane system, Golgi apparatus membrane, Lysosome membrane.
Acts as an E3 ubiquitin-protein ligase. In the presence of the human cytomegalovirus (HCMV) protein UL144, participates in 'Lys-63'-linked auto-ubiquitination of TRAF6 resulting in the virally controlled activation of NF-kappa-B at early time of infection. The C-terminus can act as an allosteric activator of the cholera toxin catalytic subunit.
Specifications
| TRIM23 | |
| Polyclonal | |
| Unconjugated | |
| TRIM23 | |
| 6330516O20Rik; ADP-ribosylation factor domain protein 1; ADP-ribosylation factor domain protein 1 64kD; ADP-ribosylation factor domain protein 1, 64kD; ADP-ribosylation factor domain protein 1, 64kDa; ADP-ribosylation factor domain-containing protein 1; AI450195; ARD1; Ard-1; ARF domain protein 1; ARFD1; E3 ubiquitin-protein ligase TRIM23; GTP-binding protein ARD-1; nucleotide binding protein; RING finger protein 46; RING-type E3 ubiquitin transferase TRIM23; RNF46; TRIM23; tripartite motif containing 23; tripartite motif protein 23; tripartite motif protein TRIM23; tripartite motif-containing 23; tripartite motif-containing protein 23 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 373, 81002, 81003 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P36406, P36407, Q8BGX0 | |
| TRIM23 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human TRIM23 (NP_0016471). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.