Learn More
Invitrogen™ TRIM23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534624
Description
Recommended positive controls: HeLaS3. Predicted reactivity: Rhesus Monkey (100%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as an E3 ubiquitin-protein ligase. In the presence of the human cytomegalovirus (HCMV) protein UL144, participates in 'Lys-63'-linked auto-ubiquitination of TRAF6 resulting in the virally controlled activation of NF-kappa-B at early time of infection. The C-terminus can act as an allosteric activator of the cholera toxin catalytic subunit.
Specifications
| TRIM23 | |
| Polyclonal | |
| Unconjugated | |
| TRIM23 | |
| 6330516O20Rik; ADP-ribosylation factor domain protein 1; ADP-ribosylation factor domain protein 1 64kD; ADP-ribosylation factor domain protein 1, 64kD; ADP-ribosylation factor domain protein 1, 64kDa; ADP-ribosylation factor domain-containing protein 1; AI450195; ARD1; Ard-1; ARF domain protein 1; ARFD1; E3 ubiquitin-protein ligase TRIM23; GTP-binding protein ARD-1; nucleotide binding protein; RING finger protein 46; RING-type E3 ubiquitin transferase TRIM23; RNF46; TRIM23; tripartite motif containing 23; tripartite motif protein 23; tripartite motif protein TRIM23; tripartite motif-containing 23; tripartite motif-containing protein 23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 373, 81003 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.67 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P36406, Q8BGX0 | |
| TRIM23 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human TRIM23. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.