missing translation for 'onlineSavingsMsg'
Learn More

TRIM23 Antibody (3E8), Novus Biologicals™

Product Code. 18356197 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356197 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356197 Supplier Novus Biologicals Supplier No. H00000373M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRIM23 Monoclonal antibody specifically detects TRIM23 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM23
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3E8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH22510
Gene Alias ADP-ribosylation factor domain protein 1, 64kDa, ADP-ribosylation factor domain-containing protein 1, ARD1GTP-binding protein ARD-1, ARF domain protein 1, ARFD1, E3 ubiquitin-protein ligase TRIM23, EC 6.3.2.-, RNF46RING finger protein 46, tripartite motif containing 23, tripartite motif protein TRIM23, tripartite motif-containing 23, Tripartite motif-containing protein 23
Host Species Mouse
Immunogen TRIM23 (AAH22510, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MATLVVNKLGAGVDSGRQGSRGTAVVKVLECGVCEDVFSLQGDKVPRLLLCGHTVCHDCLTRLPLHGRAIRCPFDRQVTDLGDSGVWGLKKNFALLELLERLQNGPIGQY
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 373
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.