missing translation for 'onlineSavingsMsg'
Learn More

Trim22 Antibody, Novus Biologicals™

Product Code. 18359229 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359229 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359229 Supplier Novus Biologicals Supplier No. H00010346B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Trim22 Polyclonal antibody specifically detects Trim22 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trim22
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_006065.2
Gene Alias 50 kDa-stimulated trans-acting factor, E3 ubiquitin-protein ligase TRIM22, EC 6.3.2, EC 6.3.2.-, GPSTAF50, RING finger protein 94, RNF94stimulated trans-acting factor (50 kDa), Staf-50, STAF50staf-50, tripartite binding motif 22, tripartite motif containing 22, tripartite motif protein TRIM22, tripartite motif-containing 22, Tripartite motif-containing protein 22
Host Species Mouse
Immunogen TRIM22 (NP_006065.2, 1 a.a. - 498 a.a.) full-length human protein. MDFSVKVDIEKEVTCPICLELLTEPLSLDCGHSFCQACITAKIKESVIISRGESSCPVCQTRFQPGNLRPNRHLANIVERVKEVKMSPQEGQKRDVCEHHGKKLQIFCKEDGKVICWVCELSQEHQGHQTFRINEVVKECQEKLQVALQRLIKEDQEAEKLEDDIRQERTAWKNYIQIERQKILKGFNEMRVILDNEEQRELQKLEEGEVNVLDNLAAATDQLVQQRQDASTLISDLQRRLRGSSVEMLQDVIDVMKRSESWTLKKPKSVSKKLKSVFRVPDLSGMLQVLKELTDVQYYWVDVMLNPGSATSNVAISVDQRQVKTVRTCTFKNSNPCDFSAFGVFGCQYFSSGKYYWEVDVSGKIAWILGVHSKISSLNKRKSSGFAFDPSVNYSKVYSRYRPQYGYWVIGLQNTCEYNAFEDSSSSDPKVLTLFMAVPPCRIGVFLDYEAGIVSFFNVTNHGALIYKFSGCRFSRPAYPYFNPWNCLVPMTVCPPSS
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10346
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.