missing translation for 'onlineSavingsMsg'
Learn More

Trim22 Antibody, Novus Biologicals™

Product Code. 18427960 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427960 25 μL 25µL
18718373 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18427960 Leverantör Novus Biologicals Leverantörsnummer NBP18179525ul

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody has been used in 5 publications

Trim22 Polyclonal specifically detects Trim22 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Trim22
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias 50 kDa-stimulated trans-acting factor, E3 ubiquitin-protein ligase TRIM22, EC 6.3.2, EC 6.3.2.-, GPSTAF50, RING finger protein 94, RNF94stimulated trans-acting factor (50 kDa), Staf-50, STAF50staf-50, tripartite binding motif 22, tripartite motif containing 22, tripartite motif protein TRIM22, tripartite motif-containing 22, Tripartite motif-containing protein 22
Gene Symbols TRIM22
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:NEVVKECQEKLQVALQRLIKEDQEAEKLEDDIRQERTAWKNYIQIERQKILKGFNEMRVILDNEEQRELQKLEEGEVNVLDNLAAATDQLVQQRQDASTLISDLQRRLRGSSVEMLQDVIDVMKRSESWTLKKPKSVSKKLKSVFRV
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10346
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.