missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM21 Recombinant Human Monoclonal Antibody (K52008_20A9)

Product Code. 30828577
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828577 50 μL
30825521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828577 Supplier Invitrogen™ Supplier No. MA564513
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Immunogen sequence: QEAREGTQGE RCAVHGERLH LFCEKDGKAL CWVCAQSRKH RDHAMVPLEE AAQEYQEKLQ VALGELRRKQ ELAEKLEVEI AIKRADWKKT VETQKSRIHA EFVQQKNFLV EEEQRQLQEL EKDEREQLRI LGEKEAKLAQ QSQALQELIS ELDRRCHSSA LELLQEVIIV LERSESWNLK Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 36%, Pig - 74%, Cynomolgus monkey - 92%.

This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The encoded protein is part of the RoSSA ribonucleoprotein, which includes a single polypeptide and one of four small RNA molecules. The RoSSA particle localizes to both the cytoplasm and the nucleus. RoSSA interacts with autoantigens in patients with Sjogren syndrome and systemic lupus erythematosus. Alternatively spliced transcript variants for this gene have been described but the full-length nature of only one has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM21
Applications ELISA
Classification Recombinant Monoclonal
Clone K52008_20A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene TRIM21
Gene Accession No. P19474
Gene Alias 52 kDa ribonucleoprotein autoantigen Ro/SS-A; 52 kDa Ro protein; 52kD Ro/SSA autoantigen; E3 ubiquitin-protein ligase TRIM21; RING finger protein 81; RING-type E3 ubiquitin transferase TRIM21; RNF81; Ro(SS-A); Ro/SSA; RO52; Sicca syndrome antigen A; sjoegren syndrome type A antigen; Sjogren syndrome antigen A1; Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro); SSA; SS-A; Ssa1; TRIM21; tripartite motif containing 21; tripartite motif protein 21; tripartite motif-containing 21; tripartite motif-containing protein 21
Gene Symbols TRIM21
Host Species Human
Immunogen Recombinant human TRIM21 (aa 81-260).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6737
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.