missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585247
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Chicken (100%), Bovine (100%). Positive Control: TRIM2 transfected 293T cell. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic filaments. Its function has not been identified.
Specifications
| TRIM2 | |
| Polyclonal | |
| Unconjugated | |
| Trim2 | |
| CMT2R; E3 ubiquitin-protein ligase TRIM2; KIAA0517; mKIAA0517; narf; Neural activity-related RING finger protein; RING finger protein 86; RING-type E3 ubiquitin transferase TRIM2; RNF86; TRIM2; tripartite motif containing 2; tripartite motif protein 2; tripartite motif protein TRIM2; tripartite motif-containing 2; tripartite motif-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23321 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.28 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9C040 | |
| Trim2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TRIM2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction