missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13663-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
| TRIM17 | |
| Polyclonal | |
| Unconjugated | |
| TRIM17 | |
| RBCC, RING finger protein 16, RNF16, terf, Testis RING finger protein, TRIM17, tripartite motif containing 17 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51127, 56631, 64702 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TPM3, Q9WV59, Q9Y577 | |
| TRIM17 | |
| TRIM17 Fusion Protein Ag4572 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction