Learn More
TRIM17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13663-1-AP-20UL
 View more versions of this product
Product Code. 16802004
  • £135.00 / 20µL
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
TRIM17
Polyclonal
Unconjugated
TRIM17
RBCC, RING finger protein 16, RNF16, terf, Testis RING finger protein, TRIM17, tripartite motif containing 17
Rabbit
Antigen Affinity Chromatography
RUO
51127, 56631, 64702
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7TPM3, Q9WV59, Q9Y577
TRIM17
TRIM17 Fusion Protein Ag4572
20 μL
Primary
Human, Rat, Mouse