missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540918
This item is not returnable.
View return policy
Description
Peptide sequence: MKEPLSRKNN VSVQCPEVAP PTRPRTVCRV PGQIEVLRGF LEDVVPDATS Sequence homology: Cow: 79%; Dog: 79%; Human: 100%; Mouse: 77%; Pig: 77%; Rabbit: 86%; Rat: 79%; Yeast: 79%.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
| TRIM17 | |
| Polyclonal | |
| Unconjugated | |
| TRIM17 | |
| E3 ubiquitin-protein ligase TRIM17; RBCC; RING finger protein 16; RING finger protein terf; RING-type E3 ubiquitin transferase TRIM17; RNF16; terf; Testis RING finger protein; Trim17; tripartite motif containing 17; tripartite motif protein 17; tripartite motif protein TRIM17; tripartite motif-containing 17; tripartite motif-containing protein 17; Unknown (protein for MGC:134518) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51127 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y577 | |
| TRIM17 | |
| Synthetic peptide directed towards the middle region of human TRIM17. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction