missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13623-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the cytoplasm. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
| TRIM15 | |
| Polyclonal | |
| Unconjugated | |
| TRIM15 | |
| RING finger protein 93, RNF93, TRIM15, tripartite motif containing 15, Zinc finger protein 178, Zinc finger protein B7, ZNF178, ZNFB7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 69097, 89870 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9C019 | |
| TRIM15 | |
| TRIM15 Fusion Protein Ag4533 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction