missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM13, Mouse anti-Human, Clone: 2A11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010206-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MELLEEDLTCPICCSLFDDPRVLPCSHNFCKKCLEGILEGSVRNSLWRPAPFKCPTCRKETSATGINSLQVNYSLKGIVEKYNKIKISPKMPVCKGHLGQSpecifications
| TRIM13 | |
| Monoclonal | |
| Unconjugated | |
| tripartite motif-containing 13 | |
| CAR/DLEU5/LEU5/RFP2/RNF77 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2A11 | |
| In 1x PBS, pH 7.4 | |
| NM_005798 | |
| TRIM13 | |
| RFP2 (NP_005789, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10206 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction