missing translation for 'onlineSavingsMsg'
Learn More

TRIM13 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18387303 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387303 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387303 Supplier Novus Biologicals Supplier No. NBP299022AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIM13 Polyclonal specifically detects TRIM13 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM13
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias B-cell chronic lymphocytic leukemia tumor suppressor Leu5, CLL-associated RING finger, E3 ubiquitin-protein ligase TRIM13, EC 6.3.2.-, LEU5CAR, Leukemia-associated protein 5, Putative tumor suppressor RFP2, Ret finger protein 2RING finger protein 77, RFP2DLEU5, RNF77Leu5, tripartite motif containing 13, tripartite motif protein 13, tripartite motif-containing 13, Tripartite motif-containing protein 13
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human TRIM13.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10206
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.