missing translation for 'onlineSavingsMsg'
Learn More

TRIM Antibody (TRIM-04), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18424894 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424894 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424894 Supplier Novus Biologicals Supplier No. NB500485MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRIM Monoclonal antibody specifically detects TRIM in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM
Applications Flow Cytometry
Classification Monoclonal
Clone TRIM-04
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Flow (Intracellular)
Formulation 50mM Sodium Borate
Gene Alias pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062
Host Species Mouse
Immunogen Recombinant fragment, corresponding to amino acids 29-186 of Human TRIM.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 50852
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.