Learn More
TRIM Antibody (488806), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB4708C
397.80 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18647953
  • £463.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
TRIM Monoclonal antibody specifically detects TRIM in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRIM
Monoclonal
PerCP
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry (Intracellular Staining)
488806
Western Blot, Intracellular Staining by Flow Cytometry
pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062
E. coli-derived recombinant human TRIM, Thr50-Asn186, Accession # Q6PIZ9
0.1 mL
Adaptive Immunity, Immunology
50852
Store at 4°C in the dark.
IgG1