missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilyl-trifluoromethanesulfonate, 97%

Product Code. 10257593 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10257593

Brand: Thermo Scientific Acros 250700500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80522-42-5 | C10H21F3O3SSi | 306.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80522-42-5
Molecular Formula C10H21F3O3SSi
Molecular Weight (g/mol) 306.42
MDL Number MFCD00009913
InChI Key LHJCZOXMCGQVDQ-UHFFFAOYSA-N
Synonym triisopropylsilyl trifluoromethanesulfonate, triisopropylsilyl triflate, triisopropylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triisopropylsilyl ester, tips-otf, tips triflate, wacker silane ip3-triflate, triisopropylsilyl trifluoromethanesulphonate, triisopropylsilyl-trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, tris 1-methylethyl silyl ester
PubChem CID 2724529
IUPAC Name tri(propan-2-yl)silyl trifluoromethanesulfonate
SMILES CC(C)[Si](C(C)C)(C(C)C)OS(=O)(=O)C(F)(F)F

Specifications

Color Brown to Brown-Orange
Density 1.1400g/mL
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CF3SO3Si(CH(CH3)2)3
Refractive Index 1.4130 to 1.415
Quantity 50 g
Fieser 13,11
Specific Gravity 1.14
Solubility Information Solubility in water: hydrolyzes
Formula Weight 306.42
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triisopropylsilyl-trifluoromethanesulfonate, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.