missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilanol, TRC

Product Code. 30425173
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425173 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425173 Supplier Toronto Research Chemicals Supplier No. TRCT219868500MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triisopropylsilanol
CAS 17877-23-5
Molecular Formula C9 H22 O Si
Quantity 500 mg
Synonym 1,1,1-Tris(1-methylethyl)silanol
InChI Formula InChI=1S/C9H22OSi/c1-7(2)11(10,8(3)4)9(5)6/h7-10H,1-6H3
SMILES CC(C)[Si](O)(C(C)C)C(C)C
IUPAC Name hydroxy-tri(propan-2-yl)silane
Molecular Weight (g/mol) 174.356
Formula Weight 174.144
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.