missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilane, TRC

Product Code. 30457045
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity unitSize
30457045 250 g 250g
1 options
This item is not returnable. View return policy

Product Code. 30457045

Brand: Toronto Research Chemicals TRCT770330250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triisopropylsilane
CAS 6485-79-6
Molecular Formula C9H22Si
Quantity 250 g
Synonym Tris(1-methylethyl)silane, Triisopropylhydrosilane
InChI Formula InChI=1S/C9H22Si/c1-7(2)10(8(3)4)9(5)6/h7-10H,1-6H3
SMILES CC(C)[SiH](C(C)C)C(C)C
IUPAC Name tri(propan-2-yl)silane
Molecular Weight (g/mol) 158.36
Formula Weight 158.1491
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.