missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl Phosphite, TRC

Product Code. 30454898
Change view
Click to view available options
Quantity:
2.5 mL
Unit Size:
2.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454898 2.5 mL 2.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454898 Supplier Toronto Research Chemicals Supplier No. TRCT8845102.5ML
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris(1-methylethoxy)phosphane
CAS 116-17-6
Molecular Formula C9 H21 O3 P
Quantity 2.5 mL
Synonym Tris(1-methylethoxy)phosphane, Phosphorous acid tris(1-methylethyl) ester, Phosphorous acid triisopropyl ester, Isopropyl phosphite, NSC 6516, Tri-2-propyl phosphite, Triisopropoxyphosphine, Triisopropyl phosphite, Tris(isopropyl) phosphite, Medronic acid Imp. A (EP)
InChI Formula InChI=1S/C9H21O3P/c1-7(2)10-13(11-8(3)4)12-9(5)6/h7-9H,1-6H3
SMILES CC(C)OP(OC(C)C)OC(C)C
IUPAC Name tripropan-2-yl phosphite
Molecular Weight (g/mol) 208.24
Formula Weight 208.1228
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.